Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRYM
Entrez Gene ID
1428
Gene Name
crystallin mu
Gene Aliases
DFNA40, THBP
Chromosome Location
Chr.16: 21258518 - 21303136 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001888.4 NP_001879.1 8 - 9 1147 62
GenBank mRNA AK290852.1 - 6 - 7 866 62
AK310424.1 - 7 - 8 958 62
BC018061.1 - 6 - 7 865 62
BX648477.1 - 8 - 9 1063 62
L02950.1 - 6 - 7 826 62
U85772.1 - 6 - 7 821 62

View Full Product Details