Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP6V0D1
Entrez Gene ID
9114
Gene Name
ATPase H+ transporting V0 subunit d1
Gene Aliases
ATP6D, ATP6DV, P39, VATX, VMA6, VPATPD
Chromosome Location
Chr.16: 67438014 - 67481186 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004691.4 NP_004682.2 2 - 3 406 77
XM_011523444.1 XP_011521746.1 3 - 4 393 77
XM_011523445.1 XP_011521747.1 3 - 4 526 77
XM_011523446.1 XP_011521748.1 2 - 3 298 77
GenBank mRNA AK301630.1 - 3 - 4 395 77
AK313035.1 - 2 - 3 364 77
BC008861.2 - 2 - 3 384 77
BI552108.1 - 2 - 3 420 77
X71490.1 - 2 - 3 331 77

View Full Product Details