The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004691.4 | NP_004682.2 | 5 - 6 | 736 | 81 |
XM_011523444.1 | XP_011521746.1 | 6 - 7 | 723 | 81 | |
XM_011523445.1 | XP_011521747.1 | 6 - 7 | 856 | 81 | |
XM_011523446.1 | XP_011521748.1 | 5 - 6 | 628 | 81 | |
GenBank mRNA | AK128641.1 | - | 1 - 2 | 2456 | 81 |
AK300570.1 | - | 6 - 7 | 791 | 81 | |
AK301630.1 | - | 6 - 7 | 725 | 81 | |
AK313035.1 | - | 5 - 6 | 694 | 81 | |
BC008861.2 | - | 5 - 6 | 714 | 81 | |
BI552108.1 | - | 5 - 6 | 749 | 80 | |
X71490.1 | - | 5 - 6 | 661 | 81 |