Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLAT
Entrez Gene ID
5327
Gene Name
plasminogen activator, tissue type
Gene Aliases
T-PA, TPA
Chromosome Location
Chr.8: 42174718 - 42207724 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000930.4 NP_000921.1 5 - 6 614 73
NM_033011.3 NP_127509.1 4 - 5 476 73
GenBank mRNA A03776.1 - 4 - 5 391 73
A07197.1 - 4 - 5 381 73
AK289387.1 - 5 - 6 566 73
AK290575.1 - 5 - 6 566 73
AK299205.1 - 5 - 6 456 73
AK313342.1 - 4 - 5 317 73
AY221101.1 - 4 - 5 357 73
BC002795.2 - 4 - 5 302 73
BC007231.1 - 5 - 6 438 73
BC013968.2 - 5 - 6 440 73
BC018636.2 - 5 - 6 440 73
BC095403.1 - 5 - 6 455 73
BT007060.1 - 4 - 5 357 73
BX641021.1 - 1 - 2 577 73
D01096.1 - 3 - 4 269 73
FM956488.1 - 4 - 5 368 73
M15518.1 - 5 - 6 433 73
M18182.1 - 4 - 5 370 73
X02901.1 - 4 - 5 374 73
X07393.1 - 5 - 6 410 73
X13097.1 - 5 - 6 465 73

View Full Product Details