The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081442.2 | NP_001074911.1 | 2 - 3 | 187 | 158 |
NM_001081443.2 | NP_001074912.1 | 2 - 3 | 187 | 158 | |
NM_001304457.1 | NP_001291386.1 | 2 - 3 | 187 | 158 | |
NM_006840.4 | NP_006831.1 | 2 - 3 | 187 | 158 | |
XM_011526359.2 | XP_011524661.1 | 2 - 3 | 171 | 158 | |
XM_011526360.2 | XP_011524662.1 | 2 - 3 | 171 | 158 | |
XM_011526361.2 | XP_011524663.1 | 2 - 3 | 171 | 158 | |
XM_011526362.2 | XP_011524664.1 | 2 - 3 | 152 | 158 | |
GenBank mRNA | AF025534.1 | - | 2 - 3 | 138 | 158 |
AK096122.1 | - | 2 - 3 | 153 | 158 | |
AK301112.1 | - | 2 - 3 | 150 | 158 | |
AK301338.1 | - | 2 - 3 | 164 | 158 | |
BC025704.1 | - | 2 - 3 | 141 | 158 | |
DC320482.1 | - | 2 - 3 | 187 | 158 |