The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081442.2 | NP_001074911.1 | 6 - 7 | 1370 | 82 |
NM_001081443.2 | NP_001074912.1 | 5 - 6 | 1070 | 82 | |
NM_001304457.1 | NP_001291386.1 | 6 - 7 | 1343 | 82 | |
NM_006840.4 | NP_006831.1 | 6 - 7 | 1370 | 82 | |
XM_011526359.2 | XP_011524661.1 | 6 - 7 | 1354 | 82 | |
XM_011526360.2 | XP_011524662.1 | 6 - 7 | 1354 | 82 | |
XM_011526361.2 | XP_011524663.1 | 6 - 7 | 1354 | 82 | |
XM_011526362.2 | XP_011524664.1 | 5 - 6 | 1035 | 82 | |
GenBank mRNA | AF025534.1 | - | 6 - 7 | 1321 | 82 |
AK057072.1 | - | 5 - 6 | 1024 | 82 | |
AK092867.1 | - | 6 - 7 | 1238 | 82 | |
AK223296.1 | - | 6 - 7 | 1326 | 82 | |
AK290845.1 | - | 6 - 7 | 1354 | 82 | |
BC025704.1 | - | 6 - 7 | 1324 | 82 |