Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- IGHG3
- Entrez Gene ID
-
3502
- Gene Name
- immunoglobulin heavy constant gamma 3 (G3m marker)
- Gene Aliases
-
IgG3
- Chromosome Location
-
Chr.14: 105765914 - 105771405 on Build GRCh38
- Gene Symbol
- IGHM
- Entrez Gene ID
-
3507
- Gene Name
- immunoglobulin heavy constant mu
- Gene Aliases
-
AGM1, MU, VH
- Chromosome Location
-
Chr.14: 105851966 - 105856217 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK090464.1 |
- |
4 - 5 |
1991 |
124 |
BX248278.1 |
- |
3 - 4 |
327 |
124 |
View Full Product Details