The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270422.1 | NP_001257351.1 | 2 - 3 | 268 | 78 |
NM_001291622.1 | NP_001278551.1 | 2 - 3 | 268 | 78 | |
NM_018658.2 | NP_061128.2 | 2 - 3 | 268 | 78 | |
XM_006721887.3 | XP_006721950.1 | 2 - 3 | 260 | 159 | |
XM_017024610.1 | XP_016880099.1 | 2 - 3 | 260 | 78 | |
GenBank mRNA | AF153815.1 | - | 2 - 3 | 208 | 78 |
AF153816.1 | - | 2 - 3 | 208 | 78 | |
AK225944.1 | - | 2 - 3 | 179 | 78 | |
DA627411.1 | - | 2 - 3 | 168 | 78 | |
DA628604.1 | - | 2 - 3 | 268 | 78 |