The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003321.4 | NP_003312.3 | 7 - 8 | 1057 | 68 |
XM_017023619.1 | XP_016879108.1 | 8 - 9 | 1314 | 68 | |
GenBank mRNA | BC001633.1 | - | 7 - 8 | 963 | 68 |
BC010041.2 | - | 7 - 8 | 971 | 68 | |
L38995.1 | - | 7 - 8 | 916 | 68 | |
S75463.1 | - | 7 - 8 | 1000 | 68 | |
X84694.1 | - | 7 - 8 | 916 | 68 |