The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003321.4 | NP_003312.3 | 8 - 9 | 1209 | 62 |
XM_011545928.2 | XP_011544230.1 | 8 - 9 | 1219 | 62 | |
XM_017023619.1 | XP_016879108.1 | 9 - 10 | 1466 | 62 | |
GenBank mRNA | BC001633.1 | - | 8 - 9 | 1115 | 62 |
BC010041.2 | - | 8 - 9 | 1123 | 62 | |
L38995.1 | - | 8 - 9 | 1068 | 62 | |
S75463.1 | - | 8 - 9 | 1152 | 62 | |
X84694.1 | - | 8 - 9 | 1068 | 62 |