Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JAKMIP2
Entrez Gene ID
9832
Gene Name
janus kinase and microtubule interacting protein 2
Gene Aliases
JAMIP2, NECC1
Chromosome Location
Chr.5: 147585439 - 147782848 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270934.1 NP_001257863.1 19 - 20 2896 121
NM_001270941.1 NP_001257870.1 20 - 21 2959 121
NM_001282282.1 NP_001269211.1 19 - 20 2682 121
XM_017010097.1 XP_016865586.1 21 - 22 2953 121
XM_017010098.1 XP_016865587.1 19 - 20 2839 121
GenBank mRNA AB011127.1 - 19 - 20 2590 121
AK299724.1 - 19 - 20 2608 121
EF512550.1 - 19 - 20 2418 121

View Full Product Details