Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARHGAP12
Entrez Gene ID
94134
Gene Name
Rho GTPase activating protein 12
Gene Aliases
-
Chromosome Location
Chr.10: 31805398 - 31928876 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AL137485.1 - 1 - 2 187 74

View Full Product Details