The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004747.3 | NP_004738.3 | 26 - 27 | 5037 | 58 |
XM_005270276.4 | XP_005270333.1 | 26 - 27 | 4991 | 58 | |
XM_006718056.3 | XP_006718119.1 | 25 - 26 | 3980 | 58 | |
XM_011540341.2 | XP_011538643.1 | 26 - 27 | 5161 | 58 | |
XM_011540342.1 | XP_011538644.1 | 26 - 27 | 4739 | 58 | |
XM_011540344.2 | XP_011538646.1 | 25 - 26 | 4646 | 58 | |
XM_011540345.1 | XP_011538647.1 | 25 - 26 | 4771 | 58 | |
XM_011540347.2 | XP_011538649.1 | 22 - 23 | 4217 | 58 | |
XM_017016913.1 | XP_016872402.1 | 26 - 27 | 4700 | 58 | |
XM_017016914.1 | XP_016872403.1 | 26 - 27 | 5030 | 58 | |
XM_017016915.1 | XP_016872404.1 | 22 - 23 | 4901 | 58 | |
GenBank mRNA | AB011155.2 | - | 26 - 27 | 5067 | 58 |
AB091676.1 | - | 24 - 25 | 4502 | 58 | |
AK314771.1 | - | 10 - 11 | 1346 | 58 | |
BC073996.1 | - | 12 - 13 | 1747 | 58 | |
BC117695.2 | - | 16 - 17 | 4795 | 58 | |
BC146794.1 | - | 26 - 27 | 5067 | 58 | |
BX649131.1 | - | 21 - 22 | 3144 | 58 | |
U61843.1 | - | 11 - 12 | 1595 | 58 |