Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LDB3
Entrez Gene ID
11155
Gene Name
LIM domain binding 3
Gene Aliases
CMD1C, CMH24, CMPD3, CYPHER, LDB3Z1, LDB3Z4, LVNC3, MFM4, ORACLE, PDLIM6, ZASP
Chromosome Location
Chr.10: 86664785 - 86736072 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080114.1 NP_001073583.1 2 - 3 269 69
NM_001080115.1 NP_001073584.1 2 - 3 269 69
NM_001080116.1 NP_001073585.1 2 - 3 374 69
NM_001171610.1 NP_001165081.1 3 - 4 391 69
NM_001171611.1 NP_001165082.1 2 - 3 269 69
NM_007078.2 NP_009009.1 2 - 3 269 69
XM_005269464.4 XP_005269521.1 3 - 4 438 69
XM_005269466.4 XP_005269523.1 3 - 4 437 69
XM_005269468.4 XP_005269525.1 3 - 4 434 69
XM_011539184.2 XP_011537486.1 3 - 4 439 69
XM_011539185.2 XP_011537487.1 3 - 4 2333 69
XM_011539186.2 XP_011537488.1 3 - 4 439 69
XM_011539187.2 XP_011537489.1 3 - 4 438 69
XM_011539188.2 XP_011537490.1 3 - 4 439 69
XM_011539189.1 XP_011537491.1 3 - 4 333 69
XM_011539190.1 XP_011537492.1 2 - 3 269 69
XM_011539191.2 XP_011537493.1 3 - 4 438 69
XM_011539193.1 XP_011537495.1 3 - 4 333 69
XM_011539194.1 XP_011537496.1 3 - 4 333 69
XM_011539195.2 XP_011537497.1 3 - 4 435 69
XM_017015606.1 XP_016871095.1 3 - 4 2333 69
XM_017015608.1 XP_016871097.1 3 - 4 2333 69
XM_017015609.1 XP_016871098.1 3 - 4 333 69
GenBank mRNA AB014513.1 - 2 - 3 267 69
AF276807.1 - 2 - 3 246 69
AF276808.1 - 2 - 3 246 69
AF276809.1 - 2 - 3 246 69
AJ133766.1 - 2 - 3 325 69
AJ133767.1 - 2 - 3 246 69
AJ133768.1 - 2 - 3 246 69
AK294696.1 - 3 - 4 326 69
AK297871.1 - 3 - 4 225 69
AK300692.1 - 3 - 4 282 69
AK304760.1 - 3 - 4 333 69
AL601484.1 - 2 - 3 281 69
BC010929.2 - 2 - 3 271 69
DA898241.1 - 2 - 3 374 69
DC357269.1 - 3 - 4 391 69
KF772970.1 - 2 - 3 246 69
KU178543.1 - 2 - 3 246 69
KU178544.1 - 2 - 3 246 69
KU178545.1 - 2 - 3 246 69

View Full Product Details