The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172357.1 | NP_001165828.1 | 2 - 3 | 357 | 56 |
NM_001172410.1 | NP_001165881.1 | 2 - 3 | 357 | 56 | |
NM_001317778.1 | NP_001304707.1 | 2 - 3 | 357 | 56 | |
NM_001317780.1 | NP_001304709.1 | 2 - 3 | 357 | 56 | |
NM_003018.3 | NP_003009.2 | 2 - 3 | 357 | 56 | |
GenBank mRNA | AK058094.1 | - | 5 - 6 | 1049 | 56 |
AK298119.1 | - | 2 - 3 | 228 | 56 | |
AK315742.1 | - | 2 - 3 | 231 | 56 | |
BC005913.1 | - | 2 - 3 | 230 | 56 | |
BG535994.1 | - | 2 - 3 | 234 | 56 | |
BG569816.1 | - | 2 - 3 | 234 | 56 | |
BI524154.1 | - | 2 - 3 | 214 | 56 | |
DQ884411.1 | - | 2 - 3 | 199 | 56 | |
J03517.1 | - | 2 - 3 | 226 | 56 | |
J03553.1 | - | 2 - 3 | 373 | 56 | |
KU178329.1 | - | 2 - 3 | 199 | 56 | |
KU178330.1 | - | 2 - 3 | 199 | 56 |