The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004949.4 | NP_004940.1 | 6 - 7 | 1238 | 142 |
NM_024422.4 | NP_077740.1 | 6 - 7 | 1238 | 142 | |
XM_005258206.4 | XP_005258263.1 | 6 - 7 | 739 | 142 | |
GenBank mRNA | AK290313.1 | - | 6 - 7 | 929 | 142 |
AK297029.1 | - | 7 - 8 | 904 | 142 | |
BC063291.1 | - | 6 - 7 | 1220 | 142 | |
BX648425.1 | - | 6 - 7 | 790 | 142 | |
EF017811.1 | - | 6 - 7 | 777 | 142 | |
EF017812.1 | - | 6 - 7 | 777 | 142 | |
X56807.1 | - | 6 - 7 | 817 | 142 |