The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321299.1 | NP_001308228.1 | 2 - 3 | 855 | 75 |
NM_032387.4 | NP_115763.2 | 2 - 3 | 855 | 75 | |
XM_011525135.2 | XP_011523437.1 | 2 - 3 | 1785 | 75 | |
XM_017024962.1 | XP_016880451.1 | 2 - 3 | 1785 | 75 | |
XM_017024963.1 | XP_016880452.1 | 2 - 3 | 1785 | 75 | |
XM_017024964.1 | XP_016880453.1 | 2 - 3 | 1785 | 75 | |
XM_017024965.1 | XP_016880454.1 | 2 - 3 | 1785 | 75 | |
GenBank mRNA | AF390018.1 | - | 2 - 3 | 808 | 75 |
AJ309861.1 | - | 1 - 2 | 169 | 75 | |
AJ316534.1 | - | 2 - 3 | 804 | 75 | |
BC136664.1 | - | 2 - 3 | 805 | 75 |