The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016458.3 | NP_057542.2 | 2 - 3 | 760 | 59 |
GenBank mRNA | AK098226.1 | - | 3 - 4 | 454 | 59 |
BC003035.1 | - | 2 - 3 | 760 | 59 | |
BC009915.2 | - | 2 - 3 | 760 | 59 | |
BC015471.1 | - | 3 - 4 | 291 | 59 |