The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016458.3 | NP_057542.2 | 4 - 5 | 954 | 123 |
GenBank mRNA | AK098226.1 | - | 5 - 6 | 648 | 123 |
BC003035.1 | - | 4 - 5 | 954 | 123 | |
BC009915.2 | - | 4 - 5 | 954 | 123 | |
BC015471.1 | - | 5 - 6 | 485 | 123 |