The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243013.1 | NP_001229942.1 | 7 - 8 | 1684 | 86 |
XM_011538099.2 | XP_011536401.1 | 6 - 7 | 1232 | 86 | |
XM_017019104.1 | XP_016874593.1 | 6 - 7 | 1232 | 86 | |
GenBank mRNA | AK027624.1 | - | 7 - 8 | 1563 | 86 |
AK304295.1 | - | 7 - 8 | 1680 | 86 | |
AL833963.1 | - | 6 - 7 | 800 | 86 |