Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRNKL1
Entrez Gene ID
51340
Gene Name
crooked neck pre-mRNA splicing factor 1
Gene Aliases
CLF, CRN, Clf1, HCRN, MSTP021, SYF3
Chromosome Location
Chr.20: 20034361 - 20056046 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278627.1 NP_001265556.1 1 - 2 99 126
GenBank mRNA AF318305.1 - 1 - 2 99 126

View Full Product Details