The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271043.2 | NP_001257972.1 | 3 - 4 | 704 | 71 |
NM_001271044.2 | NP_001257973.1 | 3 - 4 | 664 | 71 | |
NM_002501.3 | NP_002492.2 | 3 - 4 | 695 | 71 | |
XM_005259917.4 | XP_005259974.1 | 3 - 4 | 982 | 71 | |
XM_005259918.4 | XP_005259975.1 | 3 - 4 | 702 | 71 | |
XM_005259919.3 | XP_005259976.1 | 3 - 4 | 819 | 71 | |
XM_005259920.3 | XP_005259977.1 | 3 - 4 | 1381 | 71 | |
XM_006722760.2 | XP_006722823.1 | 3 - 4 | 819 | 71 | |
XM_011528040.1 | XP_011526342.1 | 3 - 4 | 702 | 71 | |
XM_017026837.1 | XP_016882326.1 | 3 - 4 | 953 | 71 | |
GenBank mRNA | AK295290.1 | - | 3 - 4 | 704 | 71 |
AK297261.1 | - | - | 663 | 71 | |
BC117113.1 | - | 2 - 3 | 658 | 71 | |
BC117115.1 | - | 2 - 3 | 658 | 71 | |
BT019732.1 | - | 3 - 4 | 627 | 71 | |
BX537794.1 | - | 3 - 4 | 701 | 71 | |
FJ861276.1 | - | 2 - 3 | 603 | 71 | |
U07811.1 | - | 2 - 3 | 574 | 71 | |
U18759.1 | - | 3 - 4 | 644 | 71 | |
U18760.1 | - | 2 - 3 | 286 | 71 | |
U18761.1 | - | 2 - 3 | 710 | 71 |