Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRB10
Entrez Gene ID
2887
Gene Name
growth factor receptor bound protein 10
Gene Aliases
GRB-IR, Grb-10, IRBP, MEG1, RSS
Chromosome Location
Chr.7: 50590063 - 50793462 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001550.2 NP_001001550.1 7 - 8 953 96
NM_001001555.2 NP_001001555.1 8 - 9 1200 96
NM_005311.4 NP_005302.3 7 - 8 873 96
XM_011515304.1 XP_011513606.1 10 - 11 1312 96
XM_011515312.2 XP_011513614.1 9 - 10 1600 96
XM_011515316.1 XP_011513618.1 8 - 9 1145 96
XM_011515323.2 XP_011513625.1 5 - 6 2649 96
XM_017012029.1 XP_016867518.1 9 - 10 1532 96
XM_017012030.1 XP_016867519.1 8 - 9 1466 96
XM_017012031.1 XP_016867520.1 8 - 9 1397 96
XM_017012032.1 XP_016867521.1 14 - 15 1909 96
XM_017012033.1 XP_016867522.1 11 - 12 1447 96
XM_017012034.1 XP_016867523.1 9 - 10 1048 96
XM_017012035.1 XP_016867524.1 15 - 16 2958 96
XM_017012036.1 XP_016867525.1 16 - 17 2743 96
XM_017012037.1 XP_016867526.1 16 - 17 2580 96
XM_017012038.1 XP_016867527.1 15 - 16 2530 96
XM_017012039.1 XP_016867528.1 12 - 13 1777 96
XM_017012040.1 XP_016867529.1 12 - 13 2313 96
XM_017012041.1 XP_016867530.1 14 - 15 2434 96
XM_017012042.1 XP_016867531.1 10 - 11 1354 96
XM_017012043.1 XP_016867532.1 11 - 12 1544 96
XM_017012044.1 XP_016867533.1 7 - 8 1143 96
XM_017012045.1 XP_016867534.1 7 - 8 807 96
XM_017012046.1 XP_016867535.1 11 - 12 2159 96
XM_017012047.1 XP_016867536.1 16 - 17 3142 96
XM_017012048.1 XP_016867537.1 15 - 16 3026 96
XM_017012049.1 XP_016867538.1 16 - 17 2648 96
XM_017012050.1 XP_016867539.1 15 - 16 2598 96
XM_017012051.1 XP_016867540.1 14 - 15 2502 96
XM_017012052.1 XP_016867541.1 14 - 15 2842 96
XM_017012053.1 XP_016867542.1 15 - 16 2464 96
XM_017012054.1 XP_016867543.1 14 - 15 2414 96
XM_017012055.1 XP_016867544.1 11 - 12 2197 96
XM_017012056.1 XP_016867545.1 10 - 11 3029 96
XM_017012057.1 XP_016867546.1 13 - 14 2318 96
XM_017012058.1 XP_016867547.1 9 - 10 1189 96
XM_017012059.1 XP_016867548.1 9 - 10 1311 96
XM_017012060.1 XP_016867549.1 17 - 18 3275 96
XM_017012061.1 XP_016867550.1 8 - 9 1141 96
XM_017012062.1 XP_016867551.1 7 - 8 902 96
XM_017012063.1 XP_016867552.1 15 - 16 3007 96
XM_017012064.1 XP_016867553.1 14 - 15 2891 96
XM_017012065.1 XP_016867554.1 12 - 13 2343 96
XM_017012066.1 XP_016867555.1 10 - 11 2702 96
XM_017012067.1 XP_016867556.1 10 - 11 1205 96
XM_017012068.1 XP_016867557.1 10 - 11 2071 96
GenBank mRNA AB000731.1 - 7 - 8 848 96
AF000017.1 - 7 - 8 842 96
AF001534.1 - 7 - 8 873 96
AJ271366.1 - 10 - 11 1312 96
AK289511.1 - 9 - 10 1167 96
BC024285.1 - 8 - 9 977 96
BX649028.1 - 7 - 8 933 96
D86962.1 - 9 - 10 1605 96
U66065.1 - 7 - 8 953 96
U69276.1 - 7 - 8 955 96

View Full Product Details