Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MS4A7
Entrez Gene ID
58475
Gene Name
membrane spanning 4-domains A7
Gene Aliases
4SPAN2, CD20L4, CFFM4, MS4A8
Chromosome Location
Chr.11: 60378485 - 60395954 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021201.4 NP_067024.1 6 - 7 843 138
NM_206938.1 NP_996821.1 5 - 6 708 138
NM_206939.1 NP_996822.1 6 - 7 795 138
NM_206940.1 NP_996823.1 5 - 6 660 138
GenBank mRNA AB026043.1 - 6 - 7 783 138
AF201951.1 - 6 - 7 754 138
AF237916.1 - 6 - 7 767 138
AF309653.1 - 6 - 7 796 138
AF350501.1 - 6 - 7 754 138
AK075106.1 - 6 - 7 847 138
AK297027.1 - 5 - 6 644 138
BC020673.1 - 6 - 7 767 138

View Full Product Details