Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FADS3
Entrez Gene ID
3995
Gene Name
fatty acid desaturase 3
Gene Aliases
CYB5RP, LLCDL3
Chromosome Location
Chr.11: 61873523 - 61892224 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021727.4 NP_068373.1 7 - 8 1048 80
XM_011545023.2 XP_011543325.1 7 - 8 1048 80
XM_017017723.1 XP_016873212.1 7 - 8 1037 80
XM_017017724.1 XP_016873213.1 7 - 8 1037 80
GenBank mRNA AF084560.1 - 7 - 8 1017 80
AF134404.1 - 7 - 8 954 80
AK091608.1 - 6 - 7 1222 80
AK295473.1 - 7 - 8 979 80
BC004901.2 - 7 - 8 1037 80

View Full Product Details