The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178133.1 | NP_001171604.1 | 2 - 3 | 506 | 70 |
NM_016169.3 | NP_057253.2 | 2 - 3 | 506 | 70 | |
XM_011539858.2 | XP_011538160.1 | 2 - 3 | 506 | 70 | |
XM_011539860.2 | XP_011538162.1 | 2 - 3 | 506 | 70 | |
XM_011539861.2 | XP_011538163.1 | 2 - 3 | 506 | 70 | |
XM_011539863.2 | XP_011538165.1 | 2 - 3 | 241 | 70 | |
XM_011539864.2 | XP_011538166.1 | 2 - 3 | 506 | 70 | |
XM_017016323.1 | XP_016871812.1 | 2 - 3 | 736 | 70 | |
GenBank mRNA | AF144231.1 | - | 2 - 3 | 461 | 70 |
AF159447.1 | - | 2 - 3 | 461 | 70 | |
AF172319.1 | - | 2 - 3 | 311 | 70 | |
AF175770.1 | - | 2 - 3 | 481 | 70 | |
AF222345.1 | - | 2 - 3 | 389 | 70 | |
AK312319.1 | - | 2 - 3 | 496 | 70 | |
AY358550.1 | - | 2 - 3 | 461 | 70 | |
BC013291.2 | - | 2 - 3 | 482 | 70 | |
DA248208.1 | - | 2 - 3 | 506 | 70 |