The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152716.2 | NP_689929.2 | 12 - 13 | 1662 | 130 |
XM_005273833.4 | XP_005273890.1 | 12 - 13 | 2784 | 130 | |
XM_005273834.1 | XP_005273891.1 | 11 - 12 | 1574 | 130 | |
XM_011544811.2 | XP_011543113.1 | 8 - 9 | 997 | 130 | |
XM_017017339.1 | XP_016872828.1 | 13 - 14 | 1738 | 130 | |
GenBank mRNA | AK094193.1 | - | 9 - 10 | 1173 | 130 |
AK127649.1 | - | 1 - 2 | 104 | 130 | |
AK127943.1 | - | 11 - 12 | 1532 | 130 | |
AK293213.1 | - | 7 - 8 | 855 | 130 | |
AL831992.2 | - | 12 - 13 | 1603 | 130 | |
BC065264.1 | - | 9 - 10 | 1156 | 130 | |
BC109038.1 | - | 9 - 10 | 1117 | 130 | |
BC109039.1 | - | 9 - 10 | 1117 | 130 | |
BC111047.1 | - | 10 - 11 | 1324 | 130 |