The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152716.2 | NP_689929.2 | 4 - 5 | 568 | 127 |
XM_005273833.4 | XP_005273890.1 | 4 - 5 | 1690 | 127 | |
XM_005273834.1 | XP_005273891.1 | 4 - 5 | 570 | 127 | |
XM_017017339.1 | XP_016872828.1 | 4 - 5 | 570 | 127 | |
GenBank mRNA | AK094193.1 | - | 1 - 2 | 79 | 127 |
AK127943.1 | - | 4 - 5 | 528 | 127 | |
AK293213.1 | - | 1 - 2 | 82 | 127 | |
AL831992.2 | - | 4 - 5 | 509 | 127 | |
BC065264.1 | - | 1 - 2 | 62 | 127 | |
BC111047.1 | - | 2 - 3 | 230 | 127 |