Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CREB3L3
Entrez Gene ID
84699
Gene Name
cAMP responsive element binding protein 3 like 3
Gene Aliases
CREB-H, CREBH, HYST1481
Chromosome Location
Chr.19: 4153601 - 4173054 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271995.1 NP_001258924.1 6 - 7 962 82
NM_001271996.1 NP_001258925.1 6 - 7 959 82
NM_032607.2 NP_115996.1 6 - 7 965 82
GenBank mRNA AB050902.1 - 6 - 7 934 82
AB073612.1 - 6 - 7 923 82
AK313099.1 - 6 - 7 937 82
BC101504.1 - 6 - 7 898 82
BC101508.1 - 6 - 7 894 82
BC143609.1 - 6 - 7 892 82

View Full Product Details