Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CREB3L3
Entrez Gene ID
84699
Gene Name
cAMP responsive element binding protein 3 like 3
Gene Aliases
CREB-H, CREBH, HYST1481
Chromosome Location
Chr.19: 4153601 - 4173054 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271995.1 NP_001258924.1 8 - 9 1115 67
NM_001271996.1 NP_001258925.1 8 - 9 1112 67
NM_001271997.1 NP_001258926.1 7 - 8 1011 67
NM_032607.2 NP_115996.1 8 - 9 1118 67
GenBank mRNA AB050902.1 - 8 - 9 1087 67
AB073612.1 - 8 - 9 1076 67
AK172839.1 - 7 - 8 961 67
AK313099.1 - 8 - 9 1090 67
BC101504.1 - 8 - 9 1051 67
BC101508.1 - 8 - 9 1047 67
BC143609.1 - 8 - 9 1045 67
BC143610.1 - 7 - 8 944 67

View Full Product Details