Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THBS1
Entrez Gene ID
7057
Gene Name
thrombospondin 1
Gene Aliases
THBS, THBS-1, TSP, TSP-1, TSP1
Chromosome Location
Chr.15: 39581079 - 39598918 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003246.3 NP_003237.2 13 - 14 2323 61
XM_011521971.1 XP_011520273.1 12 - 13 2145 61
GenBank mRNA AB209912.1 - 13 - 14 2309 61
AK291639.1 - 13 - 14 2323 61
AK304754.1 - 13 - 14 2069 61
AK310820.1 - 3 - 4 858 61
BC136469.1 - 13 - 14 2297 61
BC136470.1 - 13 - 14 2297 61
X04665.1 - 13 - 14 2219 61
X14787.1 - 13 - 14 2255 61

View Full Product Details