The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003246.3 | NP_003237.2 | 18 - 19 | 3167 | 70 |
XM_011521971.1 | XP_011520273.1 | 17 - 18 | 2989 | 70 | |
GenBank mRNA | AB209912.1 | - | 18 - 19 | 3153 | 70 |
AK304754.1 | - | 18 - 19 | 2913 | 70 | |
AK310820.1 | - | 8 - 9 | 1702 | 70 | |
BC136469.1 | - | 18 - 19 | 3141 | 70 | |
BC136470.1 | - | 18 - 19 | 3141 | 70 | |
X04665.1 | - | 18 - 19 | 3063 | 70 | |
X14787.1 | - | 18 - 19 | 3099 | 70 |