Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THBS1
Entrez Gene ID
7057
Gene Name
thrombospondin 1
Gene Aliases
THBS, THBS-1, TSP, TSP-1, TSP1
Chromosome Location
Chr.15: 39581079 - 39598918 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003246.3 NP_003237.2 20 - 21 3543 67
XM_011521971.1 XP_011520273.1 19 - 20 3365 67
GenBank mRNA AB209912.1 - 20 - 21 3529 67
AK291639.1 - 20 - 21 3543 67
AK304754.1 - 20 - 21 3289 67
AK310820.1 - 10 - 11 2078 67
BC136469.1 - 20 - 21 3517 67
BC136470.1 - 20 - 21 3517 67
M99425.1 - 2 - 3 285 67
X04665.1 - 20 - 21 3439 67
X14787.1 - 20 - 21 3475 67

View Full Product Details