Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THBS1
Entrez Gene ID
7057
Gene Name
thrombospondin 1
Gene Aliases
THBS, THBS-1, TSP, TSP-1, TSP1
Chromosome Location
Chr.15: 39581079 - 39598918 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003246.3 NP_003237.2 4 - 5 881 80
XM_011521971.1 XP_011520273.1 4 - 5 877 80
GenBank mRNA AB209912.1 - 4 - 5 867 80
AK291639.1 - 4 - 5 881 80
AK304754.1 - 4 - 5 627 80
BC028145.1 - 3 - 4 736 80
BC136469.1 - 4 - 5 855 80
BC136470.1 - 4 - 5 855 80
M14326.1 - 4 - 5 822 80
M25631.1 - 4 - 5 801 80
X04665.1 - 4 - 5 777 80
X14787.1 - 4 - 5 813 80

View Full Product Details