The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003488.3 | NP_003479.1 | 1 - 2 | 205 | 74 |
GenBank mRNA | BC000729.2 | - | 1 - 2 | 137 | 74 |
X97335.1 | - | 1 - 2 | 98 | 74 |