The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005108.3 | NP_005099.2 | 6 - 7 | 615 | 151 |
XM_011534325.2 | XP_011532627.1 | 6 - 7 | 596 | 151 | |
XM_011534327.1 | XP_011532629.1 | 6 - 7 | 596 | 151 | |
XM_011534328.2 | XP_011532630.1 | 6 - 7 | 596 | 151 | |
XM_011534329.1 | XP_011532631.1 | 6 - 7 | 596 | 151 | |
XM_011534330.2 | XP_011532632.1 | 6 - 7 | 596 | 151 | |
XM_017007595.1 | XP_016863084.1 | 4 - 5 | 364 | 151 | |
XM_017007596.1 | XP_016863085.1 | 6 - 7 | 641 | 151 | |
XM_017007598.1 | XP_016863087.1 | 6 - 7 | 596 | 151 | |
XM_017007599.1 | XP_016863088.1 | 6 - 7 | 596 | 151 | |
XM_017007600.1 | XP_016863089.1 | 4 - 5 | 450 | 151 | |
GenBank mRNA | AB015046.1 | - | 6 - 7 | 635 | 151 |
AK025728.1 | - | 6 - 7 | 567 | 151 | |
AK293325.1 | - | 4 - 5 | 369 | 151 | |
AK314386.1 | - | 6 - 7 | 563 | 151 | |
BC030231.2 | - | 6 - 7 | 584 | 151 | |
BC137076.1 | - | 6 - 7 | 547 | 151 | |
BC137080.1 | - | 6 - 7 | 547 | 151 | |
BI090475.1 | - | 6 - 7 | 478 | 151 |