Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SCN5A
- Entrez Gene ID
-
6331
- Gene Name
- sodium voltage-gated channel alpha subunit 5
- Gene Aliases
-
CDCD2, CMD1E, CMPD2, HB1, HB2, HBBD, HH1, ICCD, IVF, LQT3, Nav1.5, PFHB1, SSS1, VF1
- Chromosome Location
-
Chr.3: 38548061 - 38649673 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AJ310893.1 |
- |
1 - 2 |
40 |
78 |
View Full Product Details