Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIRT6
Entrez Gene ID
51548
Gene Name
sirtuin 6
Gene Aliases
SIR2L6
Chromosome Location
Chr.19: 4174109 - 4182604 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193285.2 NP_001180214.1 3 - 4 443 115
NM_001321058.1 NP_001307987.1 2 - 3 315 115
NM_001321060.1 NP_001307989.1 3 - 4 443 115
NM_001321061.1 NP_001307990.1 2 - 3 315 115
NM_001321064.1 NP_001307993.1 3 - 4 408 115
NM_016539.3 NP_057623.2 3 - 4 443 115
GenBank mRNA AF233396.1 - 3 - 4 438 115
AK074810.1 - 3 - 4 406 115
AK225567.1 - 3 - 4 411 115
AK293348.1 - 2 - 3 276 115
AK299420.1 - 2 - 3 258 115
AK315048.1 - 3 - 4 407 115
AL559383.3 - 3 - 4 381 115
BC004218.2 - 2 - 3 255 115
BC005026.1 - 3 - 4 381 115
BC028220.1 - 3 - 4 389 115
CN364175.1 - 3 - 4 386 115
CN364177.1 - 3 - 4 446 115
CR457200.1 - 3 - 4 378 115

View Full Product Details