The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321154.1 | NP_001308083.1 | 4 - 5 | 845 | 163 |
NM_001321155.1 | NP_001308084.1 | 5 - 6 | 969 | 163 | |
NM_001321156.1 | NP_001308085.1 | 5 - 6 | 1039 | 163 | |
NM_001321157.1 | NP_001308086.1 | 5 - 6 | 872 | 163 | |
NM_001321158.1 | NP_001308087.1 | 3 - 4 | 593 | 163 | |
NM_001321159.1 | NP_001308088.1 | 3 - 4 | 690 | 163 | |
NM_001321160.1 | NP_001308089.1 | 5 - 6 | 969 | 163 | |
NM_001321161.1 | NP_001308090.1 | 3 - 4 | 593 | 163 | |
NM_001321162.1 | NP_001308091.1 | 4 - 5 | 814 | 163 | |
NM_024770.4 | NP_079046.2 | 4 - 5 | 748 | 163 | |
GenBank mRNA | AK024046.1 | - | 4 - 5 | 815 | 163 |
AK124092.1 | - | 5 - 6 | 920 | 163 | |
AK297141.1 | - | 3 - 4 | 593 | 163 | |
BC025250.1 | - | 5 - 6 | 869 | 163 |