Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CIZ1
Entrez Gene ID
25792
Gene Name
CDKN1A interacting zinc finger protein 1
Gene Aliases
LSFR1, NP94, ZNF356
Chromosome Location
Chr.9: 128166065 - 128204383 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001131015.1 NP_001124487.1 14 - 15 2181 86
NM_001131016.1 NP_001124488.1 13 - 14 2309 86
NM_001131017.1 NP_001124489.1 14 - 15 2126 86
NM_001131018.1 NP_001124490.1 13 - 14 2069 86
NM_001257975.1 NP_001244904.1 14 - 15 2479 86
NM_001257976.1 NP_001244905.1 12 - 13 2193 86
NM_012127.2 NP_036259.2 13 - 14 2349 86
XM_005251888.3 XP_005251945.2 13 - 14 2180 86
XM_005251891.3 XP_005251948.2 14 - 15 2012 86
XM_005251892.3 XP_005251949.2 14 - 15 1997 86
XM_005251893.4 XP_005251950.4 13 - 14 1935 86
XM_006717039.3 XP_006717102.2 12 - 13 2088 86
XM_011518481.1 XP_011516783.1 13 - 14 2165 86
XM_011518482.2 XP_011516784.2 12 - 13 2103 86
XM_011518483.1 XP_011516785.1 12 - 13 2220 86
XM_017014594.1 XP_016870083.1 13 - 14 2633 86
XM_017014595.1 XP_016870084.1 13 - 14 2277 86
XM_017014596.1 XP_016870085.1 14 - 15 2124 86
GenBank mRNA AB030835.1 - 13 - 14 2262 86
AF159025.1 - 12 - 13 2141 86
AF234161.1 - 11 - 12 1912 86
AK023978.1 - 14 - 15 2181 86
AK292713.1 - 13 - 14 2300 86
AK296363.1 - 14 - 15 2480 86
AK303636.1 - 12 - 13 2176 86
AK316393.1 - 11 - 12 1905 86
BC004119.2 - 14 - 15 1966 86
BC021163.2 - 13 - 14 2048 86
EF441870.1 - 6 - 7 855 86

View Full Product Details