Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CIZ1
- Entrez Gene ID
-
25792
- Gene Name
- CDKN1A interacting zinc finger protein 1
- Gene Aliases
-
LSFR1, NP94, ZNF356
- Chromosome Location
-
Chr.9: 128166065 - 128204383 on Build GRCh38
View Full Product Details