The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012228.3 | NP_036360.3 | 3 - 4 | 399 | 110 |
GenBank mRNA | AF122004.1 | - | 3 - 4 | 316 | 110 |
AF151889.1 | - | 3 - 4 | 347 | 110 | |
BC018030.1 | - | 3 - 4 | 394 | 110 | |
BC066305.1 | - | 3 - 4 | 110 | ||
BC117471.1 | - | 3 - 4 | 448 | 110 | |
BC130380.1 | - | 3 - 4 | 448 | 110 |