Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPRD2
Entrez Gene ID
23248
Gene Name
regulation of nuclear pre-mRNA domain containing 2
Gene Aliases
HSPC099, KIAA0460
Chromosome Location
Chr.1: 150364111 - 150476566 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297673.1 NP_001284602.1 2 - 3 938 123
NM_001297674.1 NP_001284603.1 2 - 3 938 123
NM_015203.4 NP_056018.2 2 - 3 938 123
XM_005245033.2 XP_005245090.1 2 - 3 901 123
XM_006711240.2 XP_006711303.1 2 - 3 901 123
XM_011509344.2 XP_011507646.1 2 - 3 901 123
XM_017000763.1 XP_016856252.1 2 - 3 901 123
XM_017000764.1 XP_016856253.1 2 - 3 276 123
XM_017000765.1 XP_016856254.1 2 - 3 901 123
XM_017000766.1 XP_016856255.1 2 - 3 613 123
XM_017000767.1 XP_016856256.1 2 - 3 901 123
GenBank mRNA AB007929.2 - 2 - 3 307 123
AK056707.1 - 2 - 3 345 123
AK125864.1 - 3 - 4 586 123
AK291703.1 - 2 - 3 356 123
AK304380.1 - 2 - 3 356 123
AK308148.1 - 2 - 3 347 123
BC045623.1 - 2 - 3 339 123
BX641025.1 - 3 - 4 833 123

View Full Product Details