Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPRD2
Entrez Gene ID
23248
Gene Name
regulation of nuclear pre-mRNA domain containing 2
Gene Aliases
HSPC099, KIAA0460
Chromosome Location
Chr.1: 150364111 - 150476566 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297673.1 NP_001284602.1 5 - 6 1227 83
NM_001297674.1 NP_001284603.1 5 - 6 1227 83
NM_015203.4 NP_056018.2 6 - 7 1305 83
XM_005245033.2 XP_005245090.1 6 - 7 1268 83
XM_006711240.2 XP_006711303.1 6 - 7 1268 83
XM_011509344.2 XP_011507646.1 6 - 7 1268 83
XM_017000763.1 XP_016856252.1 5 - 6 1190 83
XM_017000764.1 XP_016856253.1 6 - 7 643 83
XM_017000765.1 XP_016856254.1 6 - 7 1268 83
XM_017000766.1 XP_016856255.1 5 - 6 902 83
XM_017000767.1 XP_016856256.1 5 - 6 1190 83
GenBank mRNA AB007929.2 - 6 - 7 674 83
AK056707.1 - 6 - 7 712 83
AK125864.1 - 6 - 7 875 83
AK291703.1 - 6 - 7 723 83
AK304380.1 - 5 - 6 645 83
AK308148.1 - 5 - 6 636 83
BC045623.1 - 5 - 6 628 83
BX641025.1 - 7 - 8 1200 83

View Full Product Details