Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RPGRIP1
- Entrez Gene ID
-
57096
- Gene Name
- retinitis pigmentosa GTPase regulator interacting protein 1
- Gene Aliases
-
CORD13, LCA6, RGI1, RGRIP, RPGRIP, RPGRIP1d
- Chromosome Location
-
Chr.14: 21287977 - 21351316 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF265667.1 |
- |
2 - 3 |
214 |
90 |
View Full Product Details