Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IGF2R
Entrez Gene ID
3482
Gene Name
insulin like growth factor 2 receptor
Gene Aliases
CD222, CI-M6PR, CIMPR, M6P-R, M6P/IGF2R, MPR 300, MPR1, MPR300, MPRI
Chromosome Location
Chr.6: 159969099 - 160111504 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000876.3 NP_000867.2 14 - 15 2050 128
GenBank mRNA AB209668.1 - 13 - 14 1673 128
J03528.1 - 14 - 15 2049 128
Y00285.1 - 14 - 15 2049 128

View Full Product Details