Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM38
Entrez Gene ID
10475
Gene Name
tripartite motif containing 38
Gene Aliases
RNF15, RORET
Chromosome Location
Chr.6: 25962689 - 25988867 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006355.4 NP_006346.1 1 - 2 162 84
XM_005248799.4 XP_005248856.1 1 - 2 49 84
XM_017010175.1 XP_016865664.1 1 - 2 51 84
GenBank mRNA AK313248.1 - 1 - 2 49 84
BX640949.1 - 1 - 2 79 84
DA603151.1 - 1 - 2 162 84

View Full Product Details