Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM38
Entrez Gene ID
10475
Gene Name
tripartite motif containing 38
Gene Aliases
RNF15, RORET
Chromosome Location
Chr.6: 25962689 - 25988867 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006355.4 NP_006346.1 2 - 3 403 125
XM_017010175.1 XP_016865664.1 2 - 3 292 125
GenBank mRNA AB209906.1 - 1 - 2 245 125
AK313248.1 - 2 - 3 290 125
BC026930.1 - 1 - 2 249 125
CR936707.1 - 1 - 2 114 125
DA603151.1 - 2 - 3 403 125

View Full Product Details