Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ELANE
Entrez Gene ID
1991
Gene Name
elastase, neutrophil expressed
Gene Aliases
ELA2, GE, HLE, HNE, NE, PMN-E, SCN1
Chromosome Location
Chr.19: 850997 - 856250 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001972.3 NP_001963.1 2 - 3 349 129
XM_011527775.1 XP_011526077.1 3 - 4 1026 129
XM_011527776.1 XP_011526078.1 3 - 4 365 129
GenBank mRNA BC074816.2 - 2 - 3 266 129
BC074817.2 - 2 - 3 266 129
D00187.1 - 1 - 2 139 129
M34379.1 - 2 - 3 267 129
X05875.1 - 2 - 3 267 129

View Full Product Details