The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288729.1 | NP_001275658.1 | 6 - 7 | 810 | 93 |
NM_001288730.1 | NP_001275659.1 | 6 - 7 | 800 | 93 | |
NM_001288731.1 | NP_001275660.1 | 6 - 7 | 783 | 93 | |
NM_004938.3 | NP_004929.2 | 6 - 7 | 1135 | 93 | |
XM_005251757.3 | XP_005251814.1 | 6 - 7 | 905 | 93 | |
GenBank mRNA | AA854055.1 | - | 3 - 4 | 293 | 93 |
AB208871.1 | - | 6 - 7 | 738 | 93 | |
AK295119.1 | - | 6 - 7 | 867 | 93 | |
AK296839.1 | - | 6 - 7 | 1080 | 93 | |
BC113660.1 | - | 5 - 6 | 673 | 93 | |
BC143733.1 | - | 5 - 6 | 673 | 93 | |
BC143759.1 | - | 5 - 6 | 673 | 93 | |
BT006935.1 | - | 5 - 6 | 600 | 93 | |
CR749834.1 | - | 6 - 7 | 783 | 93 | |
X76104.1 | - | 6 - 7 | 936 | 93 |